Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(rC)-binding protein 2 Mouse anti-Human, Mouse, Rat, Clone: 6A3.2, MilliporeSigma™

Catalog No. MAB11056MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11056MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB11056MI Supplier MilliporeSigma Supplier No. MAB11056

Mouse Monoclonal Antibody

Anti-Poly(rC)-binding protein 2 Antibody, clone 6A3.2 is an antibody against Poly(rC)-binding protein 2 for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Poly(rC)-binding protein 2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6A3.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001122383
Gene Symbols hnRNP-E2, HNRPE2, alpha-CP2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Poly(rC)-binding protein 2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.