Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyhistidine Tag Mouse anti-All, MilliporeSigma™

Catalog No. MAB3834MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3834MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3834MI Supplier MilliporeSigma Supplier No. MAB3834
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Polyhistidine Tag using this Anti-Polyhistidine Tag Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polyhistidine Tag
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 0.1% sodium azide.
Host Species Mouse
Immunogen His tagged recombinant protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.