Learn More
POMT2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human POMT2 peptide using ARM Technology.
Supplier: Abnova Corporation H00029954K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS)
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
POMT2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
POMT2 | |
POMT2 | |
A synthetic peptide of human POMT2 | |
RUO | |
29954 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
protein-O-mannosyltransferase 2 | |
POMT2 | |
DKFZp686G10254, FLJ22309 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.