Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PON3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002583 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-583 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-583 Supplier Abnova Corporation Supplier No. H00005446B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PON3 protein.

This gene is a member of the paraoxonase family and lies in a cluster on chromosome 7 with the other two family members. The encoded protein is secreted into the bloodstream and associates with high-density lipoprotein (HDL). The protein also rapidly hydrolyzes lactones and can inhibit the oxidation of low-density lipoprotein (LDL), a function that is believed to slow the initiation and progression of atherosclerosis. Alternatively spliced variants which encode different protein isoforms have been described; however, only one has been fully characterized. [provided by RefSeq

Sequence: MGKLVALVLLGVGLSLVGEMFLAFRERVNASREVEPVEPENCHLIEELENGSEDIDILPSGLAFISSGLKYPGMPNFAPDEPGKIFLMDLNEQNPRAQALEISGGFDKELFNPHGISIFIDKDNTVYLYVVNHPHMKSTVEIFKFEEQQRSLVYLKTIKHELLKSVNDIVVLGPEQFYATRDHYFTNSLLSFFEMILDLRWTYVLFYSPREVKVVAKGFCSANGITVSADQKYVYVADVAAKNIHIMEKHDNWDLTQLKVIQLGTLVDNLTVDPATGDILAGCHPNPMKLLNYNPEDPPGSEVLRIQNVLSEKPRVSTVYANNGSVLQGTSVASVYHGKILIGTVFHKTLYCEL

Specifications

Antigen PON3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PON3 protein.
Formulation PBS with no preservative; pH 7.4
Gene PON3
Gene Accession No. BC070374.1
Gene Symbols PON3
Host Species Mouse
Immunogen PON3 (AAH70374.1, 1 a.a. ∼ 354 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5446
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.