Learn More
Potassium Channel EAG-1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Potassium Channel EAG-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 |
| Gene Symbols | KCNH1; Kv10.1; EAG; MGC142269; hEAG1; eag; h-eag; eag1; EAG1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the C-terminus of rat Kv10.1 (Accession Q63472). The immunogen sequence is identical in mouse and 14/16 residues identical in human. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.