Learn More
Potassium Channel Kir2.1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Potassium Channel Kir2.1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 x |
| Gene Symbols | KCNJ2; IRK1; HIRK1; Kir2.1; HHBIRK1; SQT3; HHIRK1; KIR2.1; LQT7 |
| Host Species | Rabbit |
| Immunogen | Purified peptide from human Kir2.1 (amino acids 392-410) (Accession number P48049). |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.