Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Channel Kv1.2 Mouse anti-Rat, Clone: K14/16, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN77
Description
This Anti-Potassium Channel Kv1.2 Antibody, clone K14/16 is validated for use in immunohistochemistry, western blot, immunofluorescence for the detection of Potassium Channel Kv1.2.
Specifications
Potassium Channel Kv1.2 | |
Monoclonal | |
Unconjugated | |
NP_037102 | |
Mouse | |
Protein G | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunofluorescence, Immunohistochemistry, Western Blot | |
K14/16 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
KCNA2, HBK5, RBK2, HK4, MK2, KV1.2, NGK1, HUKIV, Kv1.2 | |
Recombinant protein corresponding to rat Kv1.2. | |
100 μg | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction