Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Channel Kv1.3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5178MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5178MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5178MI Supplier MilliporeSigma Supplier No. AB5178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Potassium Channel Kv1.3 using this Anti-Potassium Channel Kv1.3 Antibody validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Potassium Channel Kv1.3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 200 mL of sterile deionized water. Centrifuge antibody prepar
Gene Symbols KCNA3; HLK3; HGK5; KV1.3; HuKIII; HPCN3; Kv1.3; MK3; HUKIII; PCN3
Host Species Rabbit
Immunogen GST fusion protein and a C-terminal portion of human Kv1.3 protein (amino acids 471-523) (Accession P22001).
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002232.3
Target Species Human, Mouse, Rat
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.