Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Channel Kv4.2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5360200UL
Description
This Anti-Potassium Channel Kv4.2 Antibody is validated for use in western blotting for the detection of Potassium Channel Kv4.2.
Specifications
Potassium Channel Kv4.2 | |
Polyclonal | |
Affinity Purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.025% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Western Blot | |
Unconjugated | |
KCND2; Kv4.2; KV4.2; MGC119703; RK5; KIAA1044; MGC119702 | |
Highly purified peptide corresponding to residues 454-469 of rat Kv4.2 (Accession Q63881). | |
200 μL | |
NM_012281.2 | |
−20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction