Learn More
Potassium Channel SK3 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Potassium Channel SK3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.025% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). |
| Gene Symbols | KCNN3; OTTHUMP00000042458; OTTHUMP00000042459; SKCA3; hSK3; KCa2.3; SKCa3; K3; SK3 |
| Host Species | Rabbit |
| Immunogen | Highly purified peptide corresponding to residues 2-21 of human SK3 (Accession Q9UGI6). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.