Learn More
PP1Β Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | PP1β |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG provided as 0.2μm sterile filtered solution in phosphate buffered saline with 0.08% sodium azide. |
| Gene Symbols | PPP1CB; PP-1B; PPP1CD; MGC3672; EC 3.1.3.16 |
| Host Species | Rabbit |
| Immunogen | The immunogen used to the purified peptide conjugated to KLH corresponding to the C-terminus of PP1 beta catalytic subunit (amino acids 318-327). |
| Purification Method | Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.