Learn More
PP2A Rabbit anti-Human, Mouse, Pig, Rat, Xenopus, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | PP2A |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 100mM triethylamine, 100mM sodium phosphate, 0.02% sodium azide before the addition of glycerol to 30%. |
| Gene Symbols | PPP2R1A; PP2A-A-alpha; PR65A |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide of 13 amino acids (MAAADGDDSLY-Ahx-C), containing amino acids 1-11 of human PP2A, A subunit. Ahx is aminohexanoic acid. The immunizing sequence is conserved in human, mouse, rat, porcine and Xenopus. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.