Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A-B' (B56) Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071221MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-122-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-122-1MI Supplier MilliporeSigma Supplier No. 071221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PP2A-B’ (B56) using this Anti-PP2A-B’ (B56) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A-B’ (B56)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in PBS with 0.08% sodium azide.
Gene Symbols MGC2184 ; OTTHUMP00000022333; PP2A ; PR53 ; PTPA
Host Species Rabbit
Immunogen The immunogen used for the purified peptide conjugated to KLH corresponds to the sequence NH2-Val-Ser-Ser-Pro-His-Phe-Gln-Val-Ala-Glu- Arg-Ala-Leu-Tyr-COOH.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006243
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.