Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PP2A/Balpha (14-27) (PR55), Polyclonal

Catalog No. 539521100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
53-952-1100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-952-1100UL Supplier MilliporeSigma™ Supplier No. 539521100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PP2A/Bα (14-27) (PR55) Clone: in samples, and it is validated for Immunocytochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A/Bα (14-27) (PR55)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM Tris-HCl, 3mM glycine, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide (FSQVKGAVDDDVAE) corresponding to amino acids of the mammalian PP2A/Bα regulatory subunit, conjugated to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽55kDa regulatory subunit of PP2A/Bα. Exhibits some cross-reactivity to crude skeletal muscle extracts.
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.