Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07324MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-324-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-324-MI Supplier MilliporeSigma Supplier No. 07324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-PP2A Antibody, C subunit is validated for use in western blotting for the detection of PP2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, 5mg/ml BSA before the addition of glycerol to 30%
Gene Symbols PPP2CA; PP2CA; PP2A-alpha; RP-C; PP2Ac
Host Species Rabbit
Immunogen peptide corresponding to amino acids 290-304 (DPAPRRGEPHVTRRT) of human PP2A
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002715, NM_004156
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for 24 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.