Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Catalytic α Mouse, Unlabeled, Clone: 46, BD

Catalog No. BDB610556 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610556 150 μg
BDB610555 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610556 Supplier BD Biosciences Supplier No. 610556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P rotein p hosphatases of the type 2A (PP2A) are composed of two regulatory and one catalytic subunit. The different subunits are encoded by multiple genes. PP2A are specific for phosphoserine and phosphothreonine residues of proteins that are involved in the regulation of cellular growth. PP2A binds and dephosphorylates the middle T antigens of polyoma virus. This interaction may regulate the transcriptional activity of the virus. The 36 kDa catalytic subunit is rapidly tyrosine phosphorylated as a result of growth factor stimulation or cell transformation. Tyrosine phosphorylation of the catalytic subunit may inactivate the phosphatase activity of PP2A.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A Catalytic α
Applications Western Blot
Classification Monoclonal
Clone 46
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.