Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Mouse anti-Human, Rat, Mouse, Bovine, Rabbit, Xenopus, Yeast (S. cerevisiae), Clone: 1D6, MilliporeSigma™

Catalog No. 05421MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-421-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-421-MI Supplier MilliporeSigma Supplier No. 05421
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect PP2A using this Anti-PP2A Antibody, C subunit, clone 1D6 validated for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1M Tris-Glycine, pH 7.4, 0.15 M, NaCl, and 0.05% sodium azide. Frozen solution.
Gene Accession No. NP_002706
Gene Symbols PPP2CA, PP2CA, PP2A-alpha, PP2Ac, RP-C
Host Species Mouse
Immunogen A 16 residue synthetic peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of the 36 kDa catalytic subunit of human protein phosphatase 2A (PP2A). Clone 1D6.
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rabbit, Rat, S. cerevisiae, Xenopus species, Yeast
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.