Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Mouse anti-Bovine, Human, Mouse, Pig, Rat, Rabbit, Xenopus, Clone: 2G9, MilliporeSigma™

Catalog No. 05592MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-592-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-592-MI Supplier MilliporeSigma Supplier No. 05592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect PP2A using this Anti-PP2A Antibody, B subunit, clone 2G9 validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G9
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and glycerol to 30%
Gene Accession No. NP_001009552
Gene Symbols PPP2CA, PP2CA, PP2A-alpha, PP2Ac, RP-C
Host Species Mouse
Immunogen Peptide corresponding to residues 398-411 of human PP2A, B subunit
Purification Method Ascites
Quantity 200 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Pig, Rabbit, Rat, Xenopus species
Content And Storage −20°C for 24 months.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.