Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Mouse anti-Vertebrates, Clone: 4b7, MilliporeSigma™

Catalog No. 05577MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-577-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-577-MI Supplier MilliporeSigma Supplier No. 05577
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PP2A Antibody, C subunit, demethylated, clone 4b7 is an antibody against PP2A for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4b7
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_002715, NM_004156
Gene Symbols PPP2CA, PP2CA, PP2A-alpha, PP2Ac, RP-C
Host Species Mouse
Immunogen peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of human PP2A, C subunit
Purification Method Ascites
Quantity 200 μL
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C for 24 months.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.