Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Mouse anti-Human, Mouse, Yeast (S. cerevisiae), Clone: 7A6, MilliporeSigma™

Catalog No. 05545MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-545-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-545-MI Supplier MilliporeSigma Supplier No. 05545
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PP2A Antibody, C subunit, clone 7A6 detects level of PP2A and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7A6
Conjugate Unconjugated
Formulation Cell culture supernatant containing 30% glycerol and 0.05% sodium azide as a preservative.
Gene Accession No. NM_004156
Gene Symbols PPP2CA, PP2CA, PP2A-alpha, PP2Ac, RP-C
Host Species Mouse
Immunogen peptide (C-bARRTPDYFL) corresponding to amino acids 302-309 of human PP2A catalytic subunit
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse, S. cerevisiae, Yeast
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.