Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A Mouse anti-Human, Clone: H5D12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04639
Description
This Anti-PP2A Antibody, B subunit, B56 delta, clone H5D12 is validated for use in western blotting for the detection of PP2A.
Specifications
PP2A | |
Monoclonal | |
Unconjugated | |
NM_006245 | |
Mouse | |
100 μL | |
Human | |
Purified |
Western Blot | |
H5D12 | |
Purified mouse monoclonal IgG1 antibody in PBS with 0.05% NaN3. | |
PPP2R5D, B56D, MGC2134, MGC8949 | |
GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction