Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A-methylesterase/PME-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07095
Description
This Anti-PP2A-methylesterase/PME-1 Antibody is validated for use in immunoprecipitation, western blotting for the detection of PP2A-methylesterase/PME-1.
Specifications
PP2A-methylesterase/PME-1 | |
Polyclonal | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
PPME1; FLJ22226; PME-1 | |
A mixture of two nickel agarose-purified, His6-tagged, bacterial expressed human Protein Phosphatase Methylesterase-1 (PME-1) fragments (residues 225-302 and 277-387). These two fragments represent the carboxyl-terminal portion of the protein. | |
200 μg | |
NM_016147 | |
−20°C for 24 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction