Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PP2Balpha (482-494), Polyclonal

Catalog No. 539530100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
53-953-0100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-953-0100UL Supplier MilliporeSigma™ Supplier No. 539530100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PP2Bα (482-494) Clone: in samples, and it is validated for Immunocytochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2Bα (482-494)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM Tris-HCl, 3mM glycine, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide (RMPPRRDAMPSDA) corresponding to amino acids 482-494 of the mammalian PP2B catalytic subunit, conjugated to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa PP2B catalytic subunit. Antibody Target Gene Symbol: PPP3CA Target Synonym: 2900074D19Rik, AI841391, AW413465, Calcineurin, Calcineurin A Alpha, CALN, CALNA, CALNA1, CCN1, CN, CnA, CnA-alpha, CNA1, MGC106804, Pp2b Subunit A, PP2B-A ALPHA, PPP2B, PPP3CA alpha isoform Entrez Gene Name: protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.