Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP6C Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071224MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-122-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-122-4MI Supplier MilliporeSigma Supplier No. 071224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PP6C using this Anti-PP6C Antibody validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP6C
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing PBS with 0.08% sodium azide.
Gene Symbols PPP6C; PP6; PPP6; EC 3.1.3.16
Host Species Rabbit
Immunogen The immunogen used to the purified peptide conjugated to KLH corresponding to the sequence NH2-Cys-Ala-Val-Pro-Asp-Ser-Glu-Arg-Val-Ile- Pro-Pro-Arg-Thr-Thr-Thr-Pro-Tyr-COOH.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002721.3
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.