Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAPDC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112769
Description
A suggested positive control for this product is testislysate.
PPAPDC2 Polyclonal antibody specifically detects PPAPDC2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
PPAPDC2 | |
Polyclonal | |
PBS with 0.1% BSA and 0.08% sodium azide | |
Q8IY26 | |
PLPP6 | |
Synthetic peptide corresponding to human phosphatidic acid phosphatase type 2 domain containing 2 (PPAPDC2). | |
10 Miniblots | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PLPP6 | |
4932443D16Rik; bA6J24.6; FLJ46512; FLJ90191; MGC15483; PDP1; phosphatidic acid phosphatase type 2 domain containing 2; phosphatidic acid phosphatase type 2 domain-containing protein 2; Phospholipid phosphatase 6; PLPP6; polyisoprenoid diphosphate phosphatase type 1; PPAP2 domain-containing protein 2; Ppapdc2; Presqualene diphosphate phosphatase; PSDP; RP11-6J24.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
403313 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction