Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR γ Mouse anti-Human, Mouse, Clone: 1H4, MilliporeSigma™

Catalog No. MAB3872MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3872MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3872MI Supplier MilliporeSigma Supplier No. MAB3872
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PPAR γ Antibody, isoform 1and2 is a Mouse Monoclonal Antibody for detection of PPAR gamma also known as Peroxisome Proliferator Activated Receptor γ and has been validated in ELISA, immunocytochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR γ
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1H4
Conjugate Unconjugated
Formulation Ascites fluid containing no preservatives.
Gene Accession No. NM_005037.5; NM_015869.4; NM_138711.3; NM_138712.3
Gene Symbols PPARG, NR1C3, PPARG2, PPAR-gamma, PPARG1
Host Species Mouse
Immunogen Synthetic peptide from amino acids 107-121 of human PPAR gamma2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.