Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma 1 and 2 Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. MAB3632MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3632MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3632MI Supplier MilliporeSigma Supplier No. MAB3632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-PPAR gamma 1 and 2 Antibody, phospho-specific (Ser112) (mouse monoclonal antibody) validated in ELISA, western blot, immunocytochemistry to detect PPAR gamma 1 and 2 also known as Peroxisome Proliferator Activated Receptor γ 1 and 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma 1 and 2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Ascites. Liquid. Contains no preservative.
Gene Accession No. NM_005037.5; NM_015869.4; NM_138711.3; NM_138712.3
Gene Symbols PPARG, NR1C3, PPARG2, PPAR-gamma, PPARG1
Host Species Mouse
Immunogen Phosphorylated (serine 112) synthetic peptide from amino acids 107-121 of human PPAR gamma2 (identical to PPAR gamma1). No homology with PPAR alpha or PPAR beta.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.