Learn More
peroxisome proliferator-activated receptor gamma, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PPARG.
Supplier: Abnova Corporation H00005468A01
Description
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described. [provided by RefSeq
Sequence: FEFAVKFNALELDDSDLAIFIAVIILSGDRPGLLNVKPIEDIQDNLLQALELQLKLNHPESSQLFAKLLQKMTDLRQIVTEHVQLLQVIKKTETDMSLHPLLQEIYKDLYSpecifications
| peroxisome proliferator-activated receptor gamma | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PPARG. | |
| PPARG | |
| CIMT1/NR1C3/PPARG1/PPARG2/PPARgamma | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| 5468 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_138712 | |
| PPARG | |
| PPARG (NP_619726, 366 a.a. ∼ 475 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.