Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAT Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512333
Description
This antibody is predicted to react with chicken based on sequence homology.
PPAT is a member of the purine/pyrimidine phosphoribosyltransferase family. This protein is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis.
Specifications
PPAT | |
Polyclonal | |
Unconjugated | |
Q06203 | |
PPAT | |
KLH conjugated synthetic peptide between 175-205 amino acids from the central region of human PPAT | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
2 mg/mL | |
PPAT | |
Amidophosphoribosyltransferase [Precursor]; ATase; EC 2.4.2.14; Glutamine phosphoribosylpyrophosphate amidotransferase; GPAT; PPAT | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5471 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction