Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRF interacting protein, binding protein 2 (liprin beta 2), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018593 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-593 Supplier Abnova Corporation Supplier No. H00008495D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PPFIBP2 protein.

PPFIBP2, or liprin-beta-2, belongs to the liprin-beta family. See liprin-beta-1 (MIM 603141) for background on liprins.[supplied by OMIM

Sequence: MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHSAASNETYQERLARLEGDKESLILQVSVITDQVEAQGEKIRDLEVCLEGHQVKLNAAEEMLQQELLSRTSLETQKLDLMTEVSELKLKLVGMEKEQREQEEKQRKAEELLQELRHLKIKVEELENERNQYEWKLKATKAEVAQLQEQVALKDAEIERLHSQLSRTAALHSESHTERDQEIQRLKMGMETLLLANEDKDRRIEELTGLLNQYRKVKEIVMVTQGPSERTLSINEEEPEGGFSKWNATNKDPEELFKQEMPPRCSSPTVGPPPLPQKSLETRAQKKLSCSLEDLRSESVDKCMDGNQPFPVLEPKDSPFLAEHKYPTLPGKLSGATPNGEAAKSPPTICQPDATGSSLLRLRDTESGWDDTAVVNDLSSTSSGTESGPQSPLTPDGKRNPKGIKKFWGKIRRTQSGNFYTDTLGMAEFRRGGLRATAGPRLSRTRDSKGQKSDANAPFAQWSTERVCAWLEDFGLAQYVIFARQWVSSGHTLLTATPQDMEKELGIKHPLHRKKLVLAVKAINTKQEEKSALLDHIWVTRWLDDIGLPQYKDQFHESRVDGRMLQYLTVNDLLFLKVTSQLHHLSIKCAIHVLHVNKFNPHCLHRRPADESNLSPSEVVQWSNHRVMEWLRSVDLAEYAPNLRGSGVHGGLIILEPRFTGDTLAMLLNIPPQKTLLRRHLTTKFNALIGPEAEQEKREKMASPAYTPLTTTAKVRPRKLGFSHFGNIRKKKFDESTDYICPMEPSDGVSDSHRVYSGYRGLSPLDAPELDGLDQVGQIS

Specifications

Antigen PTPRF interacting protein, binding protein 2 (liprin beta 2)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PPFIBP2 protein.
Formulation PBS with no preservative; pH 7.4
Gene PPFIBP2
Gene Accession No. NM_003621.1
Gene Alias Cclp1/DKFZp781K06126/MGC42541
Gene Symbols PPFIBP2
Host Species Rabbit
Immunogen PPFIBP2 (NP_003612.1, 1 a.a. ∼ 876 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8495
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.