Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin D, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127785
Description
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyz
PA1-27785 detects PPIF from human, mouse, and rat samples.PA1-27785 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Peptidyl-prolyl cis-trans isomerase. mitochondrial.
Specifications
PPIF | |
Polyclonal | |
Unconjugated | |
P30405, Q99KR7, P29117 | |
PPIF | |
Synthetic peptide corresponding to human Peptidyl-prolyl cis-trans isomerase. mitochondrial. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
ELISA, Western Blot | |
1mg/mL | |
PPIF | |
RP11-342M3.4, CYP3, Cyp-D, FLJ90798, MGC117207 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
10105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction