Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1A Mouse anti-Human, Clone: 7F12, MilliporeSigma™

Catalog No. MABS415MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS415MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS415MI Supplier MilliporeSigma Supplier No. MABS415
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PPM1A, clone 7F12 Antibody is an antibody against PPM1A for use in western blotting, flow cytometry and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1A
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7F12
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.
Gene Symbols PPM1A, PPPM1A
Host Species Mouse
Immunogen Purified recombinant fragment of human PPM1A expressed in E. Coli
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.