Learn More
PPP1R12B, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PPP1R12B peptide using ARM Technology.
Supplier: Abnova Corporation H00004660K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Myosin light chain phosphatase (MLCP) consists of three subunits- catalytic subunit, large subunit/myosin binding subunit (MBS) and small subunit (sm-M20). This gene is a multi-functional gene which encodes both MBS and sm-M20. MLCP regulates myosins and the dephosphorylation is enhanced by the presence of MBS. The sm-M20 is suggested to play a regulatory role in muscle contraction by binding to MBS. MBS is also encoded by another gene, myosin light chain phosphatase target subunit 1. sm-M20 shows higher binding affinity to this gene product than to myosin light chain phosphatase target subunit 2-MBS even though the two MBS proteins are highly similar. Although both MBSs increase the activity of MLCP, myosin light chain phosphatase target subunit 1-MBS is a more efficient activator. There are four alternatively spliced transcript variants described; two alter the MBS coding region and two alter the sm-M20 coding region of this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PPP1R12B | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PPP1R12B | |
PPP1R12B | |
A synthetic peptide of human PPP1R12B | |
RUO | |
4660 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
protein phosphatase 1, regulatory (inhibitor) subunit 12B | |
PPP1R12B | |
MGC131980, MGC87886, MYPT2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.