Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein phosphatase 1, regulatory (inhibitor) subunit 1B, Mouse, Clone: 3G11, Abnova™

Catalog No. 89001370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-370 Supplier Abnova Corporation Supplier No. H00084152M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PPP1R1B.

Midbrain dopaminergic neurons play a critical role in multiple brain functions, and abnormal signaling through dopaminergic pathways has been implicated in several major neurologic and psychiatric disorders. One well-studied target for the actions of dopamine is DARPP32. In the densely dopamine- and glutamate-innervated rat caudate-putamen, DARPP32 is expressed in medium-sized spiny neurons (Ouimet and Greengard, 1990 [PubMed 2191086]) that also express dopamine D1 receptors (Walaas and Greengard, 1984 [PubMed 6319627]). The function of DARPP32 seems to be regulated by receptor stimulation. Both dopaminergic and glutamatergic (NMDA) receptor stimulation regulate the extent of DARPP32 phosphorylation, but in opposite directions (Halpain et al., 1990 [PubMed 2153935]). Dopamine D1 receptor stimulation enhances cAMP formation, resulting in the phosphorylation of DARPP32 (Walaas and Greengard, 1984 [PubMed 6319627]); phosphorylated DARPP32 is a potent protein phosphatase-1 (see MIM 176875) inhibitor (Hemmings et al., 1984 [PubMed 6087160]). NMDA receptor stimulation elevates intracellular calcium, which leads to activation of calcineurin and dephosphorylation of phospho-DARPP32, thereby reducing the phosphatase-1 inhibitory activity of DARPP32 (Halpain et al., 1990 [PubMed 2153935]).[supplied by OMIM

Sequence: MLFRLSEHSSPEEEASPHQRASGEGHHLKSKRPNPCAYTPPSLKAVQRIAESHLQSISNLNENQASEEEDELGELRELGYPREEDEEEEEDDEEEEEEEDSQAEVLKVIRQSAGQKTTCGQGLEGPWERPPPLDESERDGGSEDQVEDPALSEPGEEPQRPSPSEPGT

Specifications

Antigen protein phosphatase 1, regulatory (inhibitor) subunit 1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PPP1R1B.
Formulation PBS with no preservative; pH 7.4
Gene PPP1R1B
Gene Accession No. BC001519
Gene Alias DARPP-32/DARPP32/FLJ20940
Gene Symbols PPP1R1B
Host Species Mouse
Immunogen PPP1R1B (AAH01519, 1 a.a. ∼ 168 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84152
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.