Learn More
protein phosphatase 1, regulatory (inhibitor) subunit 3C (D01P), Rabbit anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PPP1R3C protein.
Supplier: Abnova Corporation H00005507D01P
Description
Protein phosphatase-1 (PP1; see MIM 176875) participates in the regulation of a wide variety of cellular functions by reversible protein phosphorylation. The ability of PP1 to regulate diverse functions resides in its capacity to interact with a variety of regulatory subunits that may target PP1 to specific subcellular locations, modulate its substrate specificity, and allow its activity to be responsive to extracellular signals. Several targeting subunits of PP1 have been identified, including PPP1R5, the glycogen-binding subunits PPP1R3 (MIM 600917) and PPP1R4, and the nuclear inhibitor of PP1 (PPP1R8; MIM 602636).[supplied by OMIM
Sequence: MSCTRMIQVLDPRPLTSSVMPVDVAMRLCLAHSPPVKSFLGPYDEFQRRHFVNKLKPLKSCLNIKHKAKSQNDWKCSHNQAKKRVVFADSKGLSLTAIHVFSDLPEEPAWDLQFDLLDLNDISSALKHHEEKNLILDFPQPSTDYLSFRSHFQKNFVCLENCSLQERTVTGTVKVKNVSFEKKVQIRITFDSWKNYTDVDCVYMKNVYGGTDSDTFSFAIDLPPVIPTEQKIEFCISYHANGQVFWDNNDGQNYRIVHVQWKPDGVQTQMAPQDCAFHQTSPKTELESTIFGSPRLASGLFPEWQSWGRMENLASYRSpecifications
protein phosphatase 1, regulatory (inhibitor) subunit 3C | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human PPP1R3C protein. | |
PPP1R3C | |
PPP1R5 | |
Rabbit | |
Affinity Purified | |
RUO | |
5507 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005398.3 | |
PPP1R3C | |
PPP1R3C (NP_005389.1, 1 a.a. ∼ 317 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.