Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R2A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535273
Description
The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
Specifications
| PPP2R2A | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| P63151 | |
| PPP2R2A | |
| KLH conjugated synthetic peptide between 43-71 amino acids from the N-terminal region of human PPP2R2A | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PPP2R2A | |
| PPP2R2A;Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; PP2A subunit B isoform B55-alpha; Serine/threonine-protein phosphatase 2A 55 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5520 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction