Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRCC, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89210042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-210-042 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-210-042 Supplier Abnova Corporation Supplier No. H00005546K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human PRCC peptide using ARM Technology.

In a subset of papillary renal cell carcinomas, a t(X;1)(p11;q21) chromosome translocation has been repeatedly reported and is thought to be the cause of the cancer. As a result of the translocation, the transcription factor TFE3 on the X chromosome becomes fused to this gene on chromosome 1. The fused gene results in the fusion of N-terminal proline-rich region of the protein encoded by this gene to the entire TFE3 protein. This protein has been shown to interact with the mitotic checkpoint protein MAD2B, which suggests that the dominant-negative effect of the fusion protein with TFE3 may lead to a mitotic checkpoint defect. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen PRCC
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description papillary renal cell carcinoma (translocation-associated)
Formulation In 1X PBS, pH 7.4
Gene PRCC
Gene Accession No. PRCC
Gene Alias MGC17178, MGC4723, RCCP1, TPRC
Gene Symbols PRCC
Host Species Rabbit
Immunogen A synthetic peptide of human PRCC
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5546
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.