Learn More
PRDM14 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB4350
Description
This Anti-PRDM14 Antibody is validated for use in western blotting for the detection of PRDM14.
Specifications
PRDM14 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
PRDM14 | |
GST-tagged recombinant protein corresponding to the N-terminus of human PRDM14. | |
100 μL | |
NP_078780 | |
2-8°C for 12 months |