Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM14 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABD121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABD121MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABD121MI Supplier MilliporeSigma Supplier No. ABD121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-PRDM14 Antibody is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of PRDM14 .
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM14
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Raw rabbit polyclonal serum
Gene Symbols PRDM14
Host Species Rabbit
Immunogen Human recombinant PRDM14.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.