Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM5 Rabbit anti-Human, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE114
Description
Use Anti-PRDM5 Antibody (Rabbit Polyclonal Antibody) validated in western blotting to detect PRDM5 also known as PR domain-containing protein 5.
Specifications
PRDM5 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chimpanzee, Human |
Western Blot | |
Unconjugated | |
PRDM5; PFM2 | |
Linear peptide corresponding to human PRDM5. | |
100 μg | |
NP_061169 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction