Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

peroxiredoxin 1, Mouse, Clone: 4B11-D10, Abnova™

Catalog No. 89000854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-854 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-854 Supplier Abnova Corporation Supplier No. H00005052M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PRDX1.

This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]

Sequence: MSSGNAKIGHPAPNFKATAVMPDGQFKDISLSDYKGKYVVFFFHPLDFTFVCPTEIIAFSDRAEEFKKLNCQVIGASVDSHFCHLAWVNTPKKQGGLGPMNIPLVSDPKRTIAQDYGVLKADEGISFRGLFIIDDKGILRQITVNDLPVGRSVDETLRLVQAFQFTDKHGEVCPAGWKPGSDTIKPDVQKSKEYFSKQK

Specifications

Antigen peroxiredoxin 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B11-D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PRDX1.
Formulation PBS with no preservative; pH 7.4
Gene PRDX1
Gene Accession No. BC007063
Gene Alias MSP23/NKEFA/PAG/PAGA/PAGB/PRX1/PRXI/TDPX2
Gene Symbols PRDX1
Host Species Mouse
Immunogen PRDX1 (AAH07063, 1 a.a. ∼ 199 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5052
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.