Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pre-B Cell Receptor Rat anti-Mouse, Biotin, Clone: SL156, BD

Catalog No. BDB551863 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551863 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551863 Supplier BD Biosciences Supplier No. 551863
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The pre-B cell receptor (pre-BCR) expressed during the early stages of B lymphocyte development is a heterodimer of immunoglobulin heavy chain (IgH) with surrogate light chain, which is an Ig-light-chain-like molecule composed of the non-covalently linked Lambda 5 (λ5) and VpreB proteins. The pre-BCR is believed to control IgH repertoire selection and proliferation of differentiating B lymphocytes. The SL156 antibody reacts with a conformational epitope of the pre-BCR in transfected X63-Ag8.653 cells but not with surrogate light chain, λ5, or VpreB in the absence of IgH. It detects pre-BCR on pre-B cell lines, but not on pro-B cell lines or IgM-positive splenocytes. It also detects pre-BCR on the surface of pre-B cells, but not on Ig light chain (IgL)-positive B lymphocytes, from the bone marrow of normal mice. It has been noted that the cell-surface expression of pre-BCR is upregulated after a one-hour incubation of bone-marrow leukocytes in tissue culture medium at 37°C. After immunization, cell-surface pre-BCR is detected on a subset of splenic IgL+ germinal-center B lymphocytes.

Host Species: Rat
Clone: SL156
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Purified soluble complex of recombinant μ IgH, λ5, and VpreB

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pre-B Cell Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone SL156
Concentration 0.5mg/mL
Conjugate Biotin
Description Pre-BCR
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified soluble complex of recombinant μ IgH, λ5, and VpreB
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.