Learn More
Pre-T Cell Receptor α Chain Mouse anti-Mouse, Unlabeled, Clone: 2F5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 552407
Description
At very early stages in the intrathymic differentiation of T lymphocytes which will bear αβ T Cell Receptors (TCR), the TCR β chain forms a heterodimer with the pre-TCR α chain (pTα) to form the pre-TCR. This pre-TCR associates with the signal-transducing CD3 complex and controls the survival and proliferation of CD4- CD8- (DN) thymocytes, plays a role in allelic exclusion of the TCR β chain, directly or indirectly regulates TCR δ chain expression, and is eventually replaced by the TCR α chain as thymocytes mature. The 2F5 antibody reacts with the PTα on the surface of CD44- CD25-/lo DN thymocytes.
Host Species: Mouse
Clone: 2F5
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant extracellular (Ig-like) domain of pTα
Flow Cytometry, Immunoprecipitation

Specifications
Pre-T Cell Receptor α Chain | |
Monoclonal | |
0.5mg/mL | |
pTα | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
2F5 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant extracellular (Ig-like) domain of pTα | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.