Learn More
Description
Specifications
Specifications
| Antigen | Presenilin-1 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | hPS1-NT |
| Conjugate | Unconjugated |
| Formulation | UnPurified mouse tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL. |
| Gene Accession No. | NM_000021.2 |
| Gene Symbols | PSEN1, PS1, FAD, PSNL1, PS-1, AD3, S182, Presenilin-1, EC 3.4.23.- [Contains: Presenilin-1 NTF subunit, Presenilin-1 CTF subunit, Presenilin-1 CTF12 (PS1-CTF12)]. |
| Host Species | Rat |
| Immunogen | A fusion protein antigen containing the N-terminus of human PS-1 (residues 21-80) fused to GST. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.