Learn More
MilliporeSigma™ anti-Presenilin-1, N-terminus, Clone: hPS1-NT, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1563/DEL
Description
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Presenilin-1, N-terminus | |
| Unconjugated | |
| A fusion protein antigen containing the N-terminus of human PS-1 (residues 21-80) fused to GST. | |
| RUO | |
| Primary | |
| Supernatant |
| Monoclonal | |
| Rat | |
| 100 μL | |
| Neuroscience | |
| Human | |
| IgG2a |