Learn More
MilliporeSigma™ anti-Prion Protein, a.a. 109-112 (PrP; CD230), Clone: 3F4, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1562/DEL
Description
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
Prion Protein, a.a. 109-112 (PrP; CD230) | |
Unconjugated | |
100 μg | |
Neuroscience | |
Hamster, Human | |
IgG2a |
Monoclonal | |
Mouse | |
RUO | |
Primary | |
Semi-purified |