Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prion Protein Mouse anti-Rat, Human, Sheep, Mouse, Clone: 2G11, MilliporeSigma™

Catalog No. MAB5542MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5542MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5542MI Supplier MilliporeSigma Supplier No. MAB5542

Mouse Monoclonal Antibody

Detect Prion Protein using this Anti-Prion Protein Antibody, clone 2G11 validated for use in immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prion Protein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2G11
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Accession No. NM_000311.3
Gene Symbols ASCR , CD230 , CJD , GSS, MGC26679, OTTHUMP00000030162 , OTTHUMP00000030163, PRIP, PRP, PrP, PrP27-30, PrP33-35C, PrPc, p27-30, prion
Host Species Mouse
Immunogen KLH conjugated synthetic linear peptide corresponding to the C-terminus.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Sheep
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.