Learn More
Prion Protein Mouse anti-Bovine, Mouse, Sheep, Elk, Mule Deer, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5424
Description
Anti-Prion Protein Antibody is an antibody against Prion Protein for use in ELISA, western blot, immunohistochemistry.
Specifications
Prion Protein | |
Monoclonal | |
Purified immunoglobulin. Liquid in PBS. Contains no preservative. | |
PRNP, CD230, PrP33-35C, PRP, ASCR, PrP27-30, GSS, CJD, PRIP, PrP, PrPc, MGC26679 | |
Recombinant PrP amino acids 23-237. | |
Primary | |
−20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
NM_000311.3; NM_001080121.1; NM_001080122.1; NM_001080123.1; NM_183079.2 | |
Mouse | |
100 μg | |
Bovine, Cervidae, Elk, Mouse, Sheep | |