Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prion Protein Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB5058MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5058MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5058MI Supplier MilliporeSigma Supplier No. AB5058
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Anti-Prion Protein Antibody, N-terminus, a.a. 78-97 detects level of Prion Protein and has been published and validated for use in ELISA, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prion Protein
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Goat serum. Liquid containing 0.01% thimerosal
Gene Symbols PRNP; CD230; PrP33-35C; PRP; ASCR; PrP27-30; GSS; CJD; PRIP; PrP; PrPc; MGC26679
Host Species Goat
Immunogen Synthetic peptide that corresponds to amino acids 79-97 of the N-terminus of the human PrP27-30.
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000311.3; NM_001080121.1; NM_001080122.1; NM_001080123.1; NM_183079.2
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.