Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRK1 Mouse, Unlabeled, Clone: 49, BD

Catalog No. BDB610686 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610686 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610686 Supplier BD Biosciences Supplier No. 610686

Mouse Monoclonal Antibody

Members of the Protein Kinase C (PKC) family of homologous serine/threonine protein kinases are involved in a number of processes such as cell growth, cell differentiation, and cytokine secretion. PKCs are activated by Ca2+, phospholipids, diacylglycerol, phorbol esters, and proteolysis. PRK1 (PKC-Related Kinase 1, also named PKN) was originally identified in human hippocampus as a novel protein kinase with sequence homology to PKC. PRK1 contains 942 amino acids with an apparent molecular weight of 120kDa. Although activated by limited proteolysis, PRK1 is not activated by Ca2+/diacylglycerol or phorbol esters. However, PRK1 is activated by phospholipids and arachidonic acid. PRK1 may regulate cytoskeletal changes since it binds to Rho-GTP and becomes phosphorylated in vivo, coincidentally with the formation of focal adhesions and stress fibers.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRK1
Applications Western Blot
Classification Monoclonal
Clone 49
Concentration 250μg/mL
Conjugate Unconjugated
Description PKN
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.